
(R)-(+)-Tetrahydro-2-furoic acid ≥98%, ee 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:87392-05-0
:
Formula:C₅H₈O₃
MDL Number:MFCD00211271
Boiling Pt:244 °C
Density:1.213
Flash Pt:139 °C (282 °F)
Storage Temperature:Ambient
UN:3265
ADR:8,III
:116.12 g/mol
:GHS05
Related Information
Frequently Bought Together