Spermine (N,N'-Bis(3-aminopropyl)-1,4-butanediamine)
Catalog # CHMCAB5822
Supplier: Chemicon
CAS Number:
Specifications
- Pk:100 µl
- Seq:1286
Specifications
About this item
:
:202.35 g/mol
EINECS:200-754-2
CAS Number:71-44-3
Formula:C₁₀H₂₆N₄
MDL Number:MFCD00008215