L-(-)-tert-Leucine (H-Tle-OH) for synthesis
Catalog # 8.18367.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Solubility:126 g/l (20 °C)
- Melting Point:300 °C
- Molar Mass:131.17 g/mol
- pH:5.9 (10 g/l, H2O, 22 °C)
Specification Test Results
About this item
:
:131.2 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:424-750-7
CAS Number:20859-02-3
Formula:C₆H₁₃NO₂
MDL Number:MFCD00064218