Order Entry
Netherlands
ContactUsLinkComponent
Organic size exclusion chromatography columns, GPC K-800 series preparative type
Organic size exclusion chromatography columns, GPC K-800 series preparative type
Supplier:  Resonac
Certificates
Organic size exclusion chromatography columns, GPC K-800 series preparative type
Supplier:  Resonac
Specifications
  • Catalog No:
  • SHOWF6700407
  • SHOWF6102306
  • SHOWF6102301
  • SHOWF6102303
  • SHOWF6102309
  • SHOWF6102305
  • SHOWF6102312
  • SHOWF6102304
  • SHOWF6102315
  • Description:
  • GPC K-G 8B
  • GPC K-2006
  • GPC K-2001
  • GPC K-2003
  • GPC K-2006M
  • GPC K-2005
  • GPC K-2002
  • GPC K-2004
  • GPC K-2002.5
  • Max. pressure:
  • 2,0 MPa (20 bar)
  • 2,0 MPa (20 bar)
  • 2,0 MPa (20 bar)
  • 2,0 MPa (20 bar)
  • 2,0 MPa (20 bar)
  • 2,0 MPa (20 bar)
  • 2,0 MPa (20 bar)
  • 2,0 MPa (20 bar)
  • Temperature limit:
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • 20 - 60 °C
  • USP code:
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21
  • L21