Order Entry
Netherlands
ContactUsLinkComponent
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®
Irritant
Environment
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Environment
1-Bromo-3-fluoro-4-(trifluoromethoxy)benzene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.0288 g/l (20 °C)
  • Molar Mass:
    259 g/mol
  • Boiling Point:
    171 °C (994.5 hPa)
  • Vapor Pressure:
    4 hPa (25 °C)
  • Flash Point:
    69 °C
  • Density:
    1.63 g/cm3 (20 °C)
  • pH:
    (H2O) neutral
  • Ignition Point:
    630 °C
  • Hill Formula:
    C7H3BrF4O

Specification Test Results

Related Information

Frequently Bought Together