Order Entry
Netherlands
ContactUsLinkComponent
5,5-Dimethyl-2-nitromethyl-1,3-dioxane for synthesis, Sigma-Aldrich®
5,5-Dimethyl-2-nitromethyl-1,3-dioxane for synthesis, Sigma-Aldrich®
Catalog # 8.14204.0001
Supplier:  Merck
CAS Number:  
5,5-Dimethyl-2-nitromethyl-1,3-dioxane for synthesis, Sigma-Aldrich®
Catalog # 8.14204.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 ml
  • Seq:
    2070

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    175.18 g/mol
  • Boiling Point:
    67 - 69 °C (1 hPa)
  • Density:
    1.14 g/cm3 (20 °C)
  • Hill Formula:
    C7H13NO4

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
CAS Number:33884-29-6
Formula:C₇H₁₃NO₄