Order Entry
Netherlands
ContactUsLinkComponent
Oxytetracycline hydrochloride, Supelco®
Oxytetracycline hydrochloride, Supelco®
Catalog # 46598-250MG
Supplier:  Merck
CAS Number:  
undefined
Oxytetracycline hydrochloride, Supelco®
Catalog # 46598-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :496.90 g/mol
EINECS:218-161-2
CAS Number:2058-46-0
Formula:C₂₂H₂₅ClN₂O₉
MDL Number:MFCD00135815