Buffer, PBS solution 20X concentrate (phosphate buffered saline)
PBS buffer solution 20X concentrate (phosphate buffered saline), Glass bottle, 1.000 mL
Catalog # CSTA9808S
Marketsource
Supplier: Cell Signaling
Buffer, PBS solution 20X concentrate (phosphate buffered saline)
Catalog # CSTA9808S
Supplier: Cell Signaling
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL
- Description:PBS buffer solution 20X concentrate (phosphate buffered saline)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



