Buffer, PBS solution 10X concentrate (phosphate buffered saline)
Catalog # SANTSC-24946
Leverancier: Santa Cruz Biotechnology
CAS-nummer:
Specificaties
- Pack type:Glass bottle
- Pk:1 L
- Description:PBS buffer solution 10X concentrate (phosphate buffered saline)