Order Entry
Netherlands
ContactUsLinkComponent
Free fat butyrometer
Free fat butyrometer
Catalogus # 614-0136
Supplier:  GERBER FUNKE
Free fat butyrometer
Catalogus # 614-0136
Supplier:  GERBER FUNKE
Specifications
  • Description:
    Butyrometer for free fat, complete with screw cap, scale 0,002 g
  • Pk:
    1
Specifications
About this item
For determining free fat content in milk and cream.