Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, Methidathion 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Methidathion 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15020000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Methidathion 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL15020000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    950-37-8
  • Description:
    Methidathion 10 µg/ml in cyclohexane
  • Formula:
    C₆H₁₁N₂O₄PS₃
  • Description:
    Methidathion

Specifications

About this item