Pesticide reference standard, Procymidone 100 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERXA16310000IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:32809-16-8
- Description:Procymidone 100 µg/ml in isooctane
- Formula:C₁₃H₁₁Cl₂NO₂
- Description:Procymidon