Order Entry
Netherlands
ContactUsLinkComponent
Permethrin (mixture of cis- and trans-isomeres)
Permethrin (mixture of cis- and trans-isomeres)
Catalog # EHERC15990000
Supplier:  EHRENSTORFER
CAS Number:  
Permethrin (mixture of cis- and trans-isomeres)
Catalog # EHERC15990000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
ADR:6.1,III
 :391.3 g/mol
EINECS:258-067-9
UN:2588
CAS Number:52645-53-1
Formula:C₂₁H₂₀Cl₂O₃