Order Entry
Netherlands
ContactUsLinkComponent
2,6-Di-O-methyl-β-cyclodextrine 96%
2,6-Di-O-methyl-β-cyclodextrine 96%
Certificates
undefined
2,6-Di-O-methyl-β-cyclodextrine 96%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥95 % Methylated beta-cyclodextrins ((on dry basis) calculated)
  • Water:
    ≤5.0 % (K.F.)
  • HPLC:
    35.0 to 55.0 % (ELSD) isomer purity Heptakis(2,6-di-O-methyl)-beta-cyclodextrin
  • TLC:
    ≤0.5 % unmodified beta-cyclodextrin
  • NMR:
    12.0 to 16.0 methyl groups per cyclodextrin ring (average degree of substitution)

Specification Test Results

Related Information

Frequently Bought Together