Choline hydrogen tartrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate
CAS Number:87-67-2
:
Formula:C₉H₁₉NO₇
MDL Number:MFCD00036332
Molecular Weight:253.25 g/mol
EINECS:201-763-4
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White powder to crystaline
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+16.5 to +17.5 deg (C=10, H2O)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together