

2-Imidazolidinone hemihydrate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:121325-67-5
:
Formula:C₃H₆N₂O·0.5H₂O
MDL Number:MFCD02662355
:95.10 g/mol (86.09 anhy)
:M-Clarity™ quality level = MQ200
EINECS:204-436-4
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:950 g/l (20 °C)
- Melting Point:58 °C
- Molar Mass:95.10 g/mol
- Bulk Density:720 kg/m3
- Boiling Point:132.6 °C (1013 hPa) (anhydrous substance)
- Vapor Pressure:- 24 hPa (20 °C)
- Flash Point:265 °C
- Density:1.3 g/cm3 (20 °C)
- pH:9 - 11 (100 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together