Order Entry
Netherlands
ContactUsLinkComponent
2-Ethoxyethyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Irritant
2-Ethoxyethyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Catalogus # 8.40085.0025
Supplier:  Merck
CAS Number:  
Irritant
2-Ethoxyethyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Catalogus # 8.40085.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Solubility:
    13 g/l (20 °C)
  • Molar Mass:
    158.19 g/mol
  • Boiling Point:
    205 °C
  • Vapor Pressure:
    1.5 hPa (20 °C)
  • Flash Point:
    74 °C
  • Density:
    0.96 g/cm3 (20 °C)
  • Ignition Point:
    209 °C

Specification Test Results

About this item

 :
 :158.1950 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:219-135-3
 :GHS07
Storage Temperature:Refrigerator
CAS Number:2370-63-0
Formula:C₈H₁₄O₃
MDL Number:MFCD00009249