
Ethylene glycol dimethacrylate (ethylene dimethacrylate) ≥98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-90-5
:
Formula:C₁₀H₁₄O₄
MDL Number:MFCD00008590
:198.21 g/mol
Boiling Pt:83…85 °C (1 mmHg)
Density:1.051
Flash Pt:>110 °C (230 °F)
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
EINECS:202-617-2
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together