Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, Fluorene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, Fluorene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20800000AL
 :  
Organic reference standard, Fluorene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
  EHERL20800000AL
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    86-73-7
  • Description:
    Fluorene 10 µg/ml in acetonitrile
  • Formula:
    C₁₃H₁₀
  • Description:
    Fluorene