Pesticide reference standard, Octachloronaphthalene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL20425800AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:2234-13-1
- Description:Octachloronaphthalene 10 µg/ml in acetonitrile
- Formula:C₁₀Cl₈
- Description:Octachloronaphthalene