Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, Octachloronaphthalene 10 µg/ml in isooctane, Dr Ehrenstorfer
Pesticide reference standard, Octachloronaphthalene 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20425800IO
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Octachloronaphthalene 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20425800IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    2234-13-1
  • Description:
    Octachloronaphthalene 10 µg/ml in isooctane
  • Formula:
    C₁₀Cl₈
  • Description:
    Octachloronaphthalene

Specifications

About this item