Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, ε-BHC 100 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, ε-BHC 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA14075000ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, ε-BHC 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA14075000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    6108-10-7
  • Description:
    ε-BHC 100 µg/ml in methanol
  • Formula:
    C₆H₆Cl₆
  • Description:
    ε-BHC

Specifications

About this item