Dichlorvos-[dimethyl-²H₆] (2,2-Dichlorovinyl dimethyl phosphate-[dimethyl-²H₆], DDVP-[dimethyl-²H₆])
Catalog # EHERC12530100
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:226.93 g/mol
CAS Number:203645-53-8
Formula:C₄HCl₂D₆O₄P