

N-Phenyl-1-naphthylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:90-30-2
:
Formula:C₁₆H₁₃N
MDL Number:MFCD00003878
Boiling Pt:226 °C (15 mmHg)
Melting Pt:58…62 °C
Flash Pt:>200 °C
EINECS:201-983-0
UN:3077
ADR:9,III
:219.29 g/mol
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Pink to red to brown Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:58 to 62 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together