Monoethyl fumarate potassium salt for synthesis
Catalog # 8.20588.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:182.22 g/mol
EINECS:269-879-8
CAS Number:68375-84-8
Formula:C₆H₇KO₄