Organic reference standard, Ketoprofen 10 µg/ml in methanol
Catalog # PROONE9281
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:22071-15-4
- Description:Ketoprofen 10 µg/ml in methanol
- Formula:C₁₆H₁₄O₃
- Description:Ketoprofen