Arecolinium bromide 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:300-08-3
:
Formula:C₈H₁₄BrNO₂
MDL Number:MFCD00039041
:236.11 g/mol
Melting Pt:171…174 °C
Storage Temperature:Ambient
EINECS:206-087-3
UN:1544
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:168 to 174 °C
- Titration Argentometric:≥98.0 %
Specification Test Results
Frequently Bought Together