
6-Methoxy-2-tetralone 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2472-22-2
:
Formula:C₁₁H₁₂O₂
MDL Number:MFCD00001729
:176.22 g/mol
Boiling Pt:114…116 °C (.2 mmHg)
Melting Pt:28…35 °C
Flash Pt:>110 °C
Storage Temperature:Freezer
EINECS:219-592-9
:GHS07
Specification Test Results
- Appearance:Yellow to light beige to orange Low melting crystalline solid
- Infrared spectrum:Conforms
- Melting point:28 to 35 °C
- GC:≥88 %
Specification Test Results
Frequently Bought Together