Order Entry
Netherlands
ContactUsLinkComponent
Pesticide reference standard, β-Endosulfan 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, β-Endosulfan 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL13122000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, β-Endosulfan 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL13122000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    33213-65-9
  • Description:
    β-Endosulfan 10 µg/ml in cyclohexane
  • Formula:
    C₉H₆Cl₆O₃S
  • Description:
    β-Endosulfan

Specifications

About this item