Pesticide reference standard, α-Endosulfan 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL13121000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:959-98-8
- Description:α-Endosulfan 10 µg/ml in acetonitrile
- Formula:C₉H₆Cl₆O₃S
- Description:α-Endosulfan