Herbicide reference standard, Dinoseb (2-sec-Butyl-4,6-dinitrophenol) 100 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERXA12810000AL
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:88-85-7
- Description:Dinoseb (2-sec-Butyl-4,6-dinitrophenol) 100 µg/ml in acetonitrile
- Formula:C₁₀H₁₂N₂O₅
- Description:Dinoseb (2-sec-Butyl-4,6-dinitrophenol)