2-(2-Naphthoxy)ethanol for synthesis
Catalog # 8.14891.0005
Leverancier: Merck
CAS-nummer:
Specificaties
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specificaties
Specificatie testresultaten
- Melting Point:75 °C
- Molar Mass:188.23 g/mol
Specificatie testresultaten
Over dit artikel
:
:188.23 g/mol
:M-Clarity™ quality level = MQ200
EINECS:202-228-8
CAS-nummer:93-20-9
Formule:C₁₂H₁₂O₂
MDL-nummer:MFCD00016809