Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Benzene-[D6] 2,000 µg/ml in methanol, Dr Ehrenstorfer
Benzene-[D6] 2,000 µg/ml in methanol, Dr Ehrenstorfer
EHERYA10535200ME
:
EHRENSTORFER
:
Benzene-[D6] 2,000 µg/ml in methanol, Dr Ehrenstorfer
EHERYA10535200ME
:
EHRENSTORFER
:
Pack type:
Glass bottle
Pk:
1 mL
CAS No.:
1076-43-3
Description:
Benzene-[D6] 2,000 µg/ml in methanol
Formula:
C₆H₆
Description:
Benzene-[D6]