Hydrochloric acid 3.7%, Technical Grade
VWRS83991.410
:
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
:
:∼100 °C (1013 hPa)
:1.02…1.05 g/cm³ (20 °C)
:Ambient
:1789
:8,III
:36.46 g/mol
:231-595-7
:GHS05
:7647-01-0
:HCl
:MFCD00011324