Order Entry
Netherlands
Orders LinkContactUsLinkComponent
UV lamp for microbiology

UV lamp for microbiology

366 nm, 4 W
Catalog # 1.13203.0001
Supplier:   Merck
UV lamp for microbiology
Catalog # 1.13203.0001
Supplier:  Merck
UV lamp for microbiology
UV lamp for microbiology

366 nm, 4 W

Price
154,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This UV lamp is designed for use with Fluorocult® culture media.

Specifications
  • Output:
    4 W
  • Wavelength:
    366 nm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.