Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
Pipettes
Microcapillaries
Capillaries for acid-base measurements
Capillaries for acid-base measurements
Capillaries for acid-base measurements
720-2302
:
SC SANGUIS COUNTING
Capillaries for acid-base measurements
720-2302
:
SC SANGUIS COUNTING
Description:
Capillary kit, 250 µl, including 2 caps, individually sterile packed
Pk:
10
Capillaires made of plastic, sterile.
With two closure caps