Temperature loggers, Thermo Button
About this item
Small and rugged, the Thermo Button data loggers can survive any harsh environment. They are suitable for temperature control, product tracking, transport, process analysis, HACCP.
- Collect and record date and time
- Programmable low and high alarm threshold with temporization parameters
- Battery life up to ten years depending on sample rate and temperature
- Resist to dust
Ø×H: 16×6 mm
Conform to IP 65.
Certifications: EN 12830 tested by CEMAGREF and LCIE, RTCA/DO-160 section 21, IATA (Lithium batteries), not tested for 'Food contact approval'.
Delivery information: A calibration certificate is available on request with extra charges, when ordering. The thermotrack PC software is in eight languages: French, English, German, Spanish, Portuguese, Slovenian, Japanese and Italian.
9 Options Available Below
Product Details & Documents
Small and rugged, the Thermo Button data loggers can survive any harsh environment. They are suitable for temperature control, product tracking, transport, process analysis, HACCP.
- Collect and record date and time
- Programmable low and high alarm threshold with temporization parameters
- Battery life up to ten years depending on sample rate and temperature
- Resist to dust
Ø×H: 16×6 mm
Conform to IP 65.
Certifications: EN 12830 tested by CEMAGREF and LCIE, RTCA/DO-160 section 21, IATA (Lithium batteries), not tested for 'Food contact approval'.
Delivery information: A calibration certificate is available on request with extra charges, when ordering. The thermotrack PC software is in eight languages: French, English, German, Spanish, Portuguese, Slovenian, Japanese and Italian.
Documents
PRGPDAL0052_TDS_EN
Datasheet
Specifications for Product Family
Specifications
- Catalog No:
- 620-3182
- 620-3186
- 620-3183
- 620-1853
- 620-3184
- 620-3177
- 620-3178
- 620-3179
- 620-3180
- Interface:
- USB
- Battery life:
- 10 years max.
- 10 years max.
- 30 to 50 cycles, sterilisation process
- Data memory - No. of readings:
- 2000
- 8000
- 8000
Specifications
Product Family Options
Product Information
- RangeAccuracyResolutionAvailabilityPrice
- 620-3182Compact logger, thermo-button, 21GCompact logger, thermo-button, 21GT: –40...+85 °CT: ±1,0 °C0,5 °CLog in for availability1 * 1 ST86,30 €Specifications:
More
Specifications
Cat. No.620-3182InterfaceBattery life10 years max.Data memory - No. of readings2000 - 620-3183Compact logger, thermo- button, 22LCompact logger, thermo- button, 22LT: –40...+85 °CT: ±0,5 °C0,1 ou 0,5 °CLog in for availability1 * 1 ST179,00 €Specifications:
More
Specifications
Cat. No.620-3183InterfaceBattery life10 years max.Data memory - No. of readings8000 - 620-3184Compact logger, thermo-button, 22TCompact logger, thermo-button, 22TT: 0...125 °CT: ±0,5 °C0,1 ou 0,5 °CLog in for availability1 * 1 ST367,00 €Specifications:
More
Specifications
Cat. No.620-3184InterfaceBattery life30 to 50 cycles, sterilisation processData memory - No. of readings8000
Software
Product Information
- RangeAccuracyResolutionAvailabilityPrice
- 620-3186Thermo Track software and USB Thermo Button reader, FR 21 Part 11Thermo Track software and USB Thermo Button reader, FR 21 Part 11Log in for availability1 * 1 Kit1 410,00 €Specifications:
More
Specifications
Cat. No.620-3186InterfaceUSBBattery lifeData memory - No. of readings
Capsule
Product Information
- RangeAccuracyResolutionAvailabilityPrice
- 620-1853Protection capsule for use in water or for sterilisation processProtection capsule for use in water or for sterilisation processLog in for availability1 * 1 ST114,00 €Specifications:
More
Specifications
Cat. No.620-1853InterfaceBattery lifeData memory - No. of readings
Holders
Product Information
- RangeAccuracyResolutionAvailabilityPrice
- 620-3177Key holder, blueSpecifications:
More
Specifications
Cat. No.620-3177InterfaceBattery lifeData memory - No. of readings - 620-3178Key holder, greenSpecifications:
More
Specifications
Cat. No.620-3178InterfaceBattery lifeData memory - No. of readings - 620-3179Key holder, yellowSpecifications:
More
Specifications
Cat. No.620-3179InterfaceBattery lifeData memory - No. of readings - 620-3180Key holder, redSpecifications:
More
Specifications
Cat. No.620-3180InterfaceBattery lifeData memory - No. of readings