Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
2,5-Dimethyl-1,3,4-thiadiazole for synthesis, Sigma-Aldrich®
Catalog # 8.15101.0025
Supplier:  Merck
CAS Number:  
Irritant
2,5-Dimethyl-1,3,4-thiadiazole for synthesis, Sigma-Aldrich®
Catalog # 8.15101.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Melting Point:
    61 - 64 °C
  • Molar Mass:
    114.17 g/mol
  • Boiling Point:
    202 °C (1013 hPa)

Specification Test Results

About this item

 :
 :114.17 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:248-473-4
 :GHS07
Storage Temperature:Ambient
CAS Number:27464-82-0
Formula:C₄H₆N₂S
MDL Number:MFCD00020722