2-(4-Chlorophenylthio)benzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107572-07-6
:
Formula:C₁₃H₉ClOS
MDL Number:MFCD00051766
:248.73 g/mol
Boiling Pt:102…104 °C (14 mmHg)
Melting Pt:71…74 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together