
Calcium thioglycolate trihydrate ≥98%
Catalog # 45557.18
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
Specifications
About this item
:
:276.35 g/mol
Merck Index:14,1711
Melting Pt:280…290 °C (dec.)
EINECS:212-402-5
:GHS07
Storage Temperature:Ambient
CAS Number:65208-41-5
:MFCD00043437
Formula:C₄H₆CaO₄S₂·3H₂O
MDL Number:MFCD00150707