Cholesteryl chloroformate for synthesis
Catalog # 8.41499.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Melting Point:117 - 119 °C
- Molar Mass:449.11 g/mol
Specification Test Results
About this item
:
ADR:8,II
:449.12 g/mol
EINECS:230-447-9
UN:3261
CAS Number:7144-08-3
Formula:C₂₈H₄₅ClO₂
MDL Number:MFCD00003633