Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Melatonin 99%

Melatonin 99%

Melatonin 99%
Certificates
Melatonin 99%

5 Options Available Below

Product Details & Documents
Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Formula:C₁₃H₁₆N₂O₂
  • MDL Number:MFCD00005655
  • Molecular Weight:232.28 g/mol
  • Boiling Pt:512.8 °C
  • Melting Pt:118 °C
  • Flash Pt:264 °C
  • Storage Temperature:Ambient
  • EINECS:200-797-7

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to cream to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    116 to 120 °C
  • Loss on drying:
    ≤0.5 % (1 g, 105 °C)
  • Heavy metals:
    ≤20 ppm
  • HPLC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 49,60 €
    49,60 €
    1 * 250 mg
       49,60 €

  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 104,00 €
    104,00 €
    1 * 1 g
       104,00 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 365,00 €
    365,00 €
    1 * 5 g
       365,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.