
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) ≥98% free radical
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14691-89-5
:
Formula:C₁₁H₂₁N₂O₂
MDL Number:MFCD00043593
:213.3 g/mol
Melting Pt:143…146 °C
EINECS:423-840-3
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together