

4-Aminopiperidine 95%
Catalog # L20127.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:100.16 g/mol
Boiling Pt:65 °C (18 mmHg)
Density:0.954
Flash Pt:46 °C (114 °F)
ADR:3,III
:GHS05
:GHS02
Storage Temperature:Ambient
CAS Number:13035-19-3
Formula:C₅H₁₂N₂
MDL Number:MFCD02179399