Order Entry
Netherlands
ContactUsLinkComponent
Bismuth standard solution, 1,000 mg/L Bi in dil. nitric acid standard for AAS and ICP
Bismuth standard solution, 1,000 mg/L Bi in dil. nitric acid standard for AAS and ICP
Catalog # CPIIS4400-100061
Supplier:  CPI
CAS Number:  
Bismuth standard solution, 1,000 mg/L Bi in dil. nitric acid standard for AAS and ICP
Catalog # CPIIS4400-100061
Supplier:  CPI
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Bismuth
  • Symbol:
    Bi

Specifications

About this item