Beryllium standard solution, 1,000 mg/L Be in dil. nitric acid standard for AAS and ICP
Catalog # CPIIS4400-100051
Supplier: CPI
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Beryllium
- Symbol:Be