Lead standard solution, 1000 mg/L Pb in dil. nitric acid standard for AAS and ICP
Catalog # CPIIS4400-1000281
Supplier: CPI
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Glass bottle
- Pk:100 mL
- Element:Lead
- Symbol:Pb