Order Entry
Netherlands
ContactUsLinkComponent
 
Barium standard solution, 1,000 mg/L Ba in dil. nitric acid standard for AAS and ICP
  CPIIS4400-100041
 :  CPI
 :  
Barium standard solution, 1,000 mg/L Ba in dil. nitric acid standard for AAS and ICP
  CPIIS4400-100041
 :  CPI
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Barium
  • Symbol:
    Ba