Bottles, wide neck, safety coated, WHEATON®
Supplier: DWK Life Sciences
Certificates
About this item
Bottles, made from clear soda-lime glass that conforms to USP Type III requirements. Wide neck for use with solid sample.
- Plastisol coating for safety
- Non-slip coating allows for greater ease and stability when handling wet or dry
- Black phenolic poly-vinyl lined cap
Bottles with screw caps come attached to help maintain cleanliness.
Frequently Bought Together