Order Entry
Netherlands
ContactUsLinkComponent
 
 
Formamidine acetate for synthesis, Sigma-Aldrich®
 :  Merck
 
 
Formamidine acetate for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    832 g/l (20 °C)
  • Melting Point:
    156 - 160 °C
  • Molar Mass:
    104.11 g/mol
  • pH:
    8 (400 g/l, H2O, 20 °C)
  • Ignition Point:
    565 °C

 

 

Frequently Bought Together